3,5-Diiodo-4-pyridone-1-acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(3,5-Diiodo-4-oxopyridin-1(4H)-yl)acetic acid1,4-Dihydro-3,5-diiodo-4-oxo-1-pyridineacetic Acid

B25646-09 B25646-03 B25646-09 B25646-03
AAAB25646-09EA 200.91 USD
AAAB25646-09 CAAAAB25646-09 AAAB25646-03 CAAAAB25646-03
3,5-Diiodo-4-pyridone-1-acetic acid 99%
3,5-Diiodo-4-pyridone-1-acetic acid
Formula: C₇H₅I₂NO₃
MW: 404.93 g/mol
Melting Pt: 244 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006185
CAS Number: 101-29-1
EINECS: 202-932-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR