(E)-α-(4-Chlorophenyl)cinnamonitrile 99%

Warning

B25644-14 B25644-06 B25644-06 B25644-14
AAAB25644-06EA 60.85 USD
AAAB25644-06 AAAB25644-14 CAAAAB25644-06 CAAAAB25644-14
(E)-α-(4-Chlorophenyl)cinnamonitrile 99%
(E)-alpha-(4-Chlorophenyl)cinnamonitrile
Formula: C₁₅H₁₀ClN
Melting Pt: 113…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00082873
CAS Number: 16610-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR