1,4-Diethoxybutane 98%

Warning

B25607-14 B25607-14
AAAB25607-14EA 57.88 USD
AAAB25607-14 CAAAAB25607-14
1,4-Diethoxybutane 98%
1,4-Diethoxybutane
Formula: C₈H₁₇O₂
MW: 146.23 g/mol
Boiling Pt: 165 °C
Density: 0.84
Flash Pt: 48 °C(115 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048581
CAS Number: 13344-00-8
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR