2-(N-Methyl-2-pyridylamino)ethanol 98%

Warning

Synonyms: 2-(Methyl(pyridin-2-yl)amino)ethanol2-[N-(2-Hydroxyethyl)-N-methylamino]pyridineN-Methyl-N-(2-pyridyl)ethanolamine

B25555-18 B25555-18 B25555-30 B25555-30
AAAB25555-18EA 44.5 USD
CAAAAB25555-18 AAAB25555-18 CAAAAB25555-30 AAAB25555-30
2-(N-Methyl-2-pyridylamino)ethanol 98%
2-(N-Methyl-2-pyridylamino)ethanol
Formula: C₈H₁₂N₂O
MW: 152.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02667633
CAS Number: 122321-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR