Methyl-1,2,5-trimethylpyrrole-3-carboxylate 97%

Warning

B25481-03 B25481-22 B25481-14 B25481-22 B25481-03 B25481-14
AAAB25481-03EA 17.99 USD
CAAAAB25481-14 CAAAAB25481-03 AAAB25481-03 AAAB25481-14 CAAAAB25481-22 AAAB25481-22
Methyl-1,2,5-trimethylpyrrole-3-carboxylate 97%
Methyl 1,2,5-trimethylpyrrole-3-carboxylate
Formula: C₉H₁₃NO₂
MW: 167.21
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00203867
CAS Number: 14186-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR