1-n-Propyl-4-piperidone 98%

Warning

Synonyms: 1-Propyl-4-piperidone

B25477-14 B25477-14
AAAB25477-14EA 45.87 USD
CAAAAB25477-14 AAAB25477-14
1-n-Propyl-4-piperidone 98%
1-n-Propyl-4-piperidone
Formula: C₈H₁₅NO
MW: 141.22 g/mol
Boiling Pt: 56 °C/1 mm
Density: 0.936
Flash Pt: 75 °C(168 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00010433
CAS Number: 23133-37-1
EINECS: 245-444-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR