3-Cyclopentene-1-carboxylic acid 95%
|
Warning
|
Synonyms:
cyclopent-3-ene-1-carboxylic acid
, Cyclopent-3-enecarboxylic acid
B25456-03
B25456-06
B25456-03
B25456-06
AAAB25456-03EA
112.26
USD
CAAAAB25456-03
AAAB25456-03
AAAB25456-06
CAAAAB25456-06
3-Cyclopentene-1-carboxylic acid 95%
3-Cyclopentene-1-carboxylic acid
Formula:
C₆H₈O₂ MW: 112.13 g/mol Boiling Pt: 215 °C Density: 1.084 Flash Pt: >110 °C(230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00101764 CAS Number: 7686-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...