Methyl-3-chloro-5-iodobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-chloro-5-iodobenzoate

B25444-06 B25444-03 B25444-03 B25444-06
AAAB25444-03EA 115.63 USD
CAAAAB25444-03 CAAAAB25444-06 AAAB25444-03 AAAB25444-06
Methyl-3-chloro-5-iodobenzoate 98%
Methyl 3-chloro-5-iodobenzoate
Formula: C₈H₆ClIO₂
MW: 296.49 g/mol
Melting Pt: 41…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00673018
CAS Number: 289039-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR