2-Bromo-4-nitrophenetole 99%

Warning

B25433-14
AAB25433-14EA 182.04 USD
AAB25433-14
2-Bromo-4-nitrophenetole 99%
2-Bromo-4-nitrophenetole
Formula: C₈H₈BrNO₃
MW: 246.06 g/mol
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD12547883
CAS Number: 58244-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR