(±)-Boc-α-phosphonoglycine trimethyl ester 95%

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate

B25421-14 B25421-03 B25421-06
AAB25421-06EA 191 USD
AAB25421-06 AAB25421-14 AAB25421-03
(±)-Boc-α-phosphonoglycine trimethyl ester 95%
(±)-Boc-α-phosphonoglycine trimethyl ester
Formula: C₁₀H₂₀NO₇P
MW: 297.24 g/mol
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD02178893
CAS Number: 89524-98-1
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR