Methyl-4-iodo-3-methylbenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 4-iodo-3-methylbenzoate
B25395-14
B25395-06
B25395-22
AAB25395-22EA
379.99
USD
AAB25395-22
AAB25395-14
AAB25395-06
Methyl-4-iodo-3-methylbenzoate 98%
Methyl 4-iodo-3-methylbenzoate
Formula:
C₉H₉IO₂ MW: 276.07 g/mol Melting Pt: 59…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683863 CAS Number: 5471-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...