1-Boc-isonipecotic acid ethyl ester 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl N-Boc-piperidine-4-carboxylate
, Ethyl N-BOC-4-piperidinecarboxylate
B25393-14
B25393-06
AAB25393-06EA
68.79
USD
AAB25393-06
AAB25393-14
1-Boc-isonipecotic acid ethyl ester 97+%
1-Boc-isonipecotic acid ethyl ester
Formula:
C₁₃H₂₃NO₄ MW: 257.33 g/mol Boiling Pt: 120…135 °C/0.5 mm Density: 1.046 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01763998 CAS Number: 142851-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...