Methyl-3,5-dibromo-4-methylbenzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 3,5-dibromo-4-methylbenzoate

B25348-14 B25348-06 B25348-14 B25348-06
AAAB25348-14EA 189 USD
CAAAAB25348-14 AAAB25348-14 CAAAAB25348-06 AAAB25348-06
Methyl-3,5-dibromo-4-methylbenzoate 98%
Methyl 3,5-dibromo-4-methylbenzoate
Formula: C₉H₈Br₂O₂
MW: 307.98 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00010729
CAS Number: 74896-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR