Ethyl-4-isopropylpiperazine-1-carboxylate 98%

Warning

Synonyms: Ethyl 4-isopropylpiperazine-1-carboxylate

B25341-06 B25341-06 B25341-03 B25341-03
AAAB25341-06EA 104.67 USD
AAAB25341-06 CAAAAB25341-06 AAAB25341-03 CAAAAB25341-03
Ethyl-4-isopropylpiperazine-1-carboxylate 98%
Ethyl-4-isopropylpiperazine-1-carboxylate
Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Boiling Pt: 128…130 °C/14 mm
Storage Temperature: Ambient
MDL Number: MFCD08064222
CAS Number: 61014-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR