Methyl-4-methyl-3-nitrobenzoate 99%

Synonyms: Methyl 4-methyl-3-nitrobenzoate

B25299-06 B25299-22 B25299-22 B25299-06 B25299-14 B25299-14
AAAB25299-22EA 579.32 USD
CAAAAB25299-22 AAAB25299-22 AAAB25299-14 CAAAAB25299-14 AAAB25299-06 CAAAAB25299-06
Methyl-4-methyl-3-nitrobenzoate 99%
Methyl 4-methyl-3-nitrobenzoate
Formula: C₉H₉NO₄
MW: 195.18 g/mol
Melting Pt: 49…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00130054
CAS Number: 7356-11-8
EINECS: 230-886-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR