1-(4-Chlorophenyl)piperazine dihydrochloride 95%
B25283-06
B25283-22
B25283-22
B25283-06
B25283-14
AAAB25283-06EA
21.85
USD
AAAB25283-06
CAAAAB25283-06
CAAAAB25283-22
AAAB25283-14
AAAB25283-22
1-(4-Chlorophenyl)piperazine dihydrochloride 95%
1-(4-Chlorophenyl)piperazine dihydrochloride
5-HT2C/2B receptor agonist/partial agonist
Formula:
C₁₀H₁₅Cl₃N₂ Melting Pt: 275…278 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012766 CAS Number: 38869-46-4 EINECS: 254-165-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...