6-Methoxyflavanone 98%
|
Warning
|
B25240-03
B25240-03
B25240-14
B25240-06
B25240-06
AAAB25240-03EA
51.13
USD
CAAAAB25240-03
CAAAAB25240-14
CAAAAB25240-06
AAAB25240-03
AAAB25240-06
6-Methoxyflavanone 98%
6-Methoxyflavanone
Formula:
C₁₆H₁₄O₃ Melting Pt: 141…143 °C Storage Temperature: Ambient |
EINECS:
000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...