2,7-Dimethyl-9-fluorenone-4-carboxylic acid 97%
|
Warning
|
B25228-14
B25228-14
B25228-03
B25228-06
B25228-03
B25228-06
AAAB25228-14EA
682.65
USD
CAAAAB25228-14
CAAAAB25228-03
AAAB25228-14
AAAB25228-03
CAAAAB25228-06
AAAB25228-06
2,7-Dimethyl-9-fluorenone-4-carboxylic acid 97%
2,7-Dimethyl-9-fluorenone-4-carboxylic acid
Formula:
C₁₆H₁₂O₃ MW: 252.27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04039780 CAS Number: 500536-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...