(2-Chlorophenoxy)acetamide 97%

Warning

Synonyms: 2-(2-Chlorophenoxy)acetamide

B25223-14 B25223-03 B25223-06 B25223-03 B25223-06 B25223-14
AAAB25223-06EA 99.56 USD
AAAB25223-06 CAAAAB25223-06 AAAB25223-14 AAAB25223-03 CAAAAB25223-03 CAAAAB25223-14
(2-Chlorophenoxy)acetamide 97%
(2-Chlorophenoxy)acetamide
Formula: C₈H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00173819
CAS Number: 35368-68-4
EINECS: 252-530-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR