2-(Tribromomethyl)quinoline 97%
|
Warning
|
Synonyms:
α,α,α-Tribromoquinaldine
B25218-09
B25218-30
B25218-18
B25218-09
B25218-18
AAAB25218-18EA
120.99
USD
CAAAAB25218-09
AAAB25218-18
CAAAAB25218-18
CAAAAB25218-30
AAAB25218-09
2-(Tribromomethyl)quinoline 97%
2-(Tribromomethyl)quinoline
Formula:
C₁₀H₆Br₃N Melting Pt: 128…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006755 CAS Number: 613-53-6 EINECS: 210-347-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...