Methylmaleic hydrazide 97%

Warning

Synonyms: 4-Methyl-3,6-pyridazinediol3,6-Dihydroxy-4-methylpyridazine1,2-Dihydro-4-methyl-3,6-pyridazinedioneMethylmaleic Acid Hydrazide

B25152-14 B25152-03 B25152-14 B25152-06 B25152-03 B25152-06
AAAB25152-06EA 121.5 USD
AAAB25152-06 CAAAAB25152-06 AAAB25152-03 AAAB25152-14 CAAAAB25152-14 CAAAAB25152-03
Methylmaleic hydrazide 97%
Methylmaleic hydrazide
Formula: C₅H₆N₂O₂
MW: 126.12 g/mol
Melting Pt: 284…288 °C
Storage Temperature: Ambient
MDL Number: MFCD00067746
CAS Number: 5754-18-7
EINECS: 227-276-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR