1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole 96%
|
Warning
|
Synonyms:
1-(3,5-Bis(trifluoromethyl)phenyl)-2,5-dimethyl-1H-pyrrole
B25087-06
B25087-03
B25087-14
B25087-03
B25087-06
B25087-14
AAAB25087-03EA
32.13
USD
CAAAAB25087-14
CAAAAB25087-03
CAAAAB25087-06
AAAB25087-03
AAAB25087-14
AAAB25087-06
1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole 96%
1-[3,5-Bis(trifluoromethyl)phenyl]-2,5-dimethylpyrrole
Formula:
C₁₄H₁₁F₆N MW: 307,23 g/mol Boiling Pt: 78…80 °C/0.1 mm Storage Temperature: Ambient |
MDL Number:
MFCD00084902 CAS Number: 175205-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...