1-(2,4,6-Triisopropylphenylsulfonyl)-1,2,4-triazole 98%

Warning

Synonyms: 1-((2,4,6-Triisopropylphenyl)sulphonyl)-1H-1,2,4-triazole1-((2,4,6-Triisopropylphenyl)sulfonyl)-1H-1,2,4-triazole

B25038-03 B25038-03 B25038-14 B25038-06 B25038-14 B25038-06
AAAB25038-03EA 23.85 USD
CAAAAB25038-03 CAAAAB25038-14 AAAB25038-03 AAAB25038-14 CAAAAB25038-06 AAAB25038-06
1-(2,4,6-Triisopropylphenylsulfonyl)-1,2,4-triazole 98%
1-(2,4,6-Triisopropylphenylsulphonyl)-1,2,4-triazole
Formula: C₁₇H₂₅N₃O₂S
MW: 335.47 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014089
CAS Number: 54230-60-3
EINECS: 259-035-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR