2-Phenylmalonamide 97%

Supplier: Thermo Scientific Chemicals
Warning

B24972-14 B24972-03 B24972-14 B24972-06 B24972-03 B24972-06
AAAB24972-14EA 539.99 USD
CAAAAB24972-14 CAAAAB24972-03 AAAB24972-14 AAAB24972-03 CAAAAB24972-06 AAAB24972-06
2-Phenylmalonamide 97%
2-Phenylmalonamide
Formula: C₉H₁₀N₂O₂
Melting Pt: ca 233 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051776
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR