2-Butyl-5-methylthiophene 97%

B24943-06 B24943-03 B24943-14 B24943-14 B24943-06 B24943-03
AAAB24943-06EA 74.32 USD
AAAB24943-06 CAAAAB24943-06 AAAB24943-03 AAAB24943-14 CAAAAB24943-14 CAAAAB24943-03
2-Butyl-5-methylthiophene 97%
2-Butyl-5-methylthiophene
Formula: C₉H₁₄S
Storage Temperature: Ambient
MDL Number: MFCD00091566
CAS Number: 111510-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR