1-Bromo-4-heptyloxybenzene 97%

B24844-03 B24844-03 B24844-06 B24844-06
AAAB24844-03EA 43.75 USD
AAAB24844-03 CAAAAB24844-03 CAAAAB24844-06 AAAB24844-06
1-Bromo-4-heptyloxybenzene 97%
1-Bromo-4-heptyloxybenzene
Formula: C₁₃H₁₉BrO
Storage Temperature: Ambient
MDL Number: MFCD00173752
CAS Number: 123732-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR