1-Chloro-2,5-dipropoxybenzene 97%

B24776-30 B24776-30 B24776-18 B24776-18 B24776-09 B24776-09
AAAB24776-18EA 115.44 USD
AAAB24776-18 CAAAAB24776-09 AAAB24776-09 CAAAAB24776-30 AAAB24776-30 CAAAAB24776-18
1-Chloro-2,5-dipropoxybenzene 97%
1-Chloro-2,5-dipropoxybenzene
Formula: C₁₂H₁₇ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00048688
CAS Number: 916791-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR