1,4-Dibutoxy-2-nitrobenzene 98%

Warning

B24709-06 B24709-06 B24709-22 B24709-14 B24709-14 B24709-22
AAAB24709-06EA 44.24 USD
CAAAAB24709-22 AAAB24709-06 CAAAAB24709-06 AAAB24709-14 CAAAAB24709-14 AAAB24709-22
1,4-Dibutoxy-2-nitrobenzene 98%
1,4-Dibutoxy-2-nitrobenzene
Formula: C₁₄H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00272266
CAS Number: 135-15-9
EINECS: 205-180-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR