2,2'-Thenoin 97%

Supplier: Thermo Scientific Chemicals

Warning

B24671-03 B24671-03 B24671-14 B24671-06 B24671-06 B24671-14
AAB24671-06EA 185.3 USD
CAAAB24671-03 CAAAB24671-14 AAB24671-06 AAB24671-03 CAAAB24671-06 AAB24671-14
2,2'-Thenoin 97%
2,2'-Thenoin
Formula: C₁₀H₈O₂S₂
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00022505
CAS Number: 27761-02-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR