1-(4-Nitrophenylazo)-2-naphthol

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Para Red1-((4-Nitrophenyl)diazenyl)naphthalen-2-ol

B24600-14 B24600-22 B24600-22 B24600-14
AAB24600-22EA 177.99 USD
AAB24600-22 CAAAB24600-14 AAB24600-14 CAAAB24600-22
1-(4-Nitrophenylazo)-2-naphthol
1-(4-Nitrophenylazo)-2-naphthol
Formula: C₁₆H₁₁N₃O₃
MW: 293.28 g/mol
Melting Pt: 248…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00003908
CAS Number: 6410-10-2
EINECS: 229-093-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR