4-Fluoro-3-(trifluoromethyl)benzophenone 97+%

Warning

B24599-03 B24599-03 B24599-06 B24599-06
AAB24599-03EA 50.76 USD
AAB24599-03 CAAAB24599-06 AAB24599-06 CAAAB24599-03
4-Fluoro-3-(trifluoromethyl)benzophenone 97+%
4-Fluoro-3-(trifluoromethyl)benzophenone
Formula: C₁₄H₈F₄O
Melting Pt: 44…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00061258
CAS Number: 239087-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR