5-Chloro-2-benzoxazolinone 99%

Warning

Synonyms: 5-Chloro-2(3H)-benzoxazoloneChloroxazone5-Chlorobenzo[d]oxazol-2(3H)-one

B24507-30 B24507-18 B24507-18 B24507-0B B24507-30 B24507-0B
AAB24507-18EA 62.79 USD
CAAAB24507-18 AAB24507-18 CAAAB24507-0B AAB24507-30 CAAAB24507-30 AAB24507-0B
5-Chloro-2-benzoxazolinone 99%
5-Chloro-2-benzoxazolinone
Formula: C₇H₄ClNO₂
MW: 169.57 g/mol
Melting Pt: 189…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00005717
CAS Number: 95-25-0
EINECS: 202-403-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR