Benzyltrimethylammonium tribromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Mono(N,N,N-trimethyl-1-phenylmethanaminium) tribromide
B24470-06
B24470-14
B24470-14
B24470-06
B24470-03
B24470-03
AAB24470-14EA
228
USD
CAAAB24470-14
CAAAB24470-03
AAB24470-14
AAB24470-03
CAAAB24470-06
AAB24470-06
Benzyltrimethylammonium tribromide 98%
Benzyltrimethylammonium tribromide
Formula:
C₁₀H₁₆Br₃N MW: 389.97 g/mol Melting Pt: 99…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134423 CAS Number: 111865-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...