Beryllon II

Warning

Synonyms: 2-(8-Hydroxy-3,6-disulfo-1-naphthylazo)-1,8-dihydroxynaphthalene-3,6-disulfonic acid tetrasodium salt8-(1,8-Dihydroxy-3,6-disulpho-2-naphthylazo)-1-naphthol-3,6-disulphonic acid tetrasodium salt2-(8-Hydroxy-3,6-disulpho-1-naphthylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic acid tetrasodium salt8-(1,8-Dihydroxy-3,6-disulfo-2-naphthylazo)-1-naphthol-3,6-disulfonic acid tetrasodium salt

B24387-06 B24387-06
AAB24387-06EA 52.49 USD
CAAAB24387-06 AAB24387-06
Beryllon II
Beryllon II
Formula: C₂₀H₁₄N₂O₁₅S₄
MW: 738.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046409
CAS Number: 51550-25-5
EINECS: 257-273-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR