4-(Benzyloxy)phenylboronic acid 97%

Warning

Synonyms: (4-(Benzyloxy)phenyl)boronic acid4-Benzyloxybenzeneboronic acid

B24351-01 B24351-06 B24351-03
AAB24351-01EA 16.76 USD
AAB24351-01 AAB24351-03 AAB24351-06
4-(Benzyloxy)phenylboronic acid 97%
4-(Benzyloxy)phenylboronic acid
Formula: C₁₃H₁₃BO₃
MW: 228.06 g/mol
Melting Pt: 193…198 °C
Storage Temperature: Ambient
MDL Number: MFCD01075705
CAS Number: 146631-00-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR