2,3-Difluoro-4-methylbenzamide 97+%

Warning

B24254-06 B24254-06 B24254-03 B24254-03
AAB24254-03EA 36.51 USD
CAAAB24254-03 AAB24254-03 AAB24254-06 CAAAB24254-06
2,3-Difluoro-4-methylbenzamide 97+%
2,3-Difluoro-4-methylbenzamide
Formula: C₈H₇F₂NO
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD01631633
CAS Number: 261763-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR