(3,4,5-Trimethoxyphenyl)acetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4,5-Trimethoxyphenylacetonitrile

B24247-14 B24247-14
AAB24247-14EA 233.99 USD
CAAAB24247-14 AAB24247-14
(3,4,5-Trimethoxyphenyl)acetonitrile 97%
(3,4,5-Trimethoxyphenyl)acetonitrile
Formula: C₁₁H₁₃NO₃
MW: 207.23 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00001912
EINECS: 236-388-5
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR