4-Methyl-3-(trifluoromethyl)benzoic acid 97+%

Warning

B24246-03 B24246-06
AAB24246-06EA 259.74 USD
AAB24246-06 AAB24246-03
4-Methyl-3-(trifluoromethyl)benzoic acid 97+%
4-Methyl-3-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Melting Pt: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD01631592
CAS Number: 261952-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR