Hexadecacarbonylhexarhodium 98%

Warning

Synonyms: Hexarhodium hexadecacarbonyl

B24185-02 B24185-77 B24185-01 B24185-01 B24185-02 B24185-77
AAAB24185-77EA 154.99 USD
CAAAAB24185-01 CAAAAB24185-77 AAAB24185-77 AAAB24185-01 CAAAAB24185-02 AAAB24185-02
Hexadecacarbonylhexarhodium 98%
Hexadecacarbonylhexarhodium
Formula: C₁₆O₁₆Rh₆
MW: 1065.595 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
EINECS: 249-009-3
UN: 3466
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR