Ethyl-3-piperidinecarboxylate 97%

Danger

Synonyms: Ethyl piperidine-3-carboxylateEthyl nipecotate

B24155-22 B24155-22 B24155-14 B24155-14
AAB24155-14EA 37.18 USD
CAAAB24155-22 CAAAB24155-14 AAB24155-14 AAB24155-22
Ethyl-3-piperidinecarboxylate 97%
Ethyl 3-piperidinecarboxylate
Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 102…104 °C/7 mm
Density: 1.012
Flash Pt: 90 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005991
CAS Number: 5006-62-2
EINECS: 225-681-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR