1-Butyl-4-chlorobenzene 97%

B24138-06 B24138-22 B24138-22 B24138-06 B24138-14 B24138-14
AAB24138-06EA 40.82 USD
AAB24138-06 CAAAB24138-06 CAAAB24138-14 CAAAB24138-22 AAB24138-22 AAB24138-14
1-Butyl-4-chlorobenzene 97%
1-Butyl-4-chlorobenzene
Formula: C₁₀H₁₃Cl
MW: 168.67
Boiling Pt: 225…228 °C
Density: 1,00
Storage Temperature: Ambient
MDL Number: MFCD00051926
CAS Number: 15499-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR