2-Piperazinone 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: Piperazin-2-one

B24131-06 B24131-06 B24131-03 B24131-03
AAB24131-06EA 246 USD
CAAAB24131-03 AAB24131-06 AAB24131-03 CAAAB24131-06
2-Piperazinone 97%
2-Piperazinone
Formula: C₄H₈N₂O
MW: 100.12 g/mol
Melting Pt: 129…134 °C
Storage Temperature: Ambient
MDL Number: MFCD01318687
CAS Number: 5625-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR