1-(2-Pyrimidyl)piperazine dihydrochloride ≥98%

Warning

Synonyms: 2-(1-Piperazinyl)pyrimidine Dihydrochloride

B24085-06 B24085-22 B24085-06 B24085-14 B24085-14 B24085-22
AAB24085-22EA 277.58 USD
CAAAB24085-22 AAB24085-22 AAB24085-14 CAAAB24085-06 AAB24085-06 CAAAB24085-14
1-(2-Pyrimidyl)piperazine dihydrochloride ≥98%
1-(2-Pyrimidyl)piperazine dihydrochloride
Formula: C₈H₁₂N₄
MW: 237.13 g/mol
Melting Pt: ca 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012779
CAS Number: 94021-22-4
EINECS: 301-415-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR