3-Chloro-2,6-difluorobenzamide 97+%

Warning

B24065-06 B24065-06 B24065-03 B24065-03
AAB24065-03EA 37.6 USD
CAAAB24065-06 AAB24065-03 AAB24065-06 CAAAB24065-03
3-Chloro-2,6-difluorobenzamide 97+%
3-Chloro-2,6-difluorobenzamide
Formula: C₇H₄ClF₂NO
Melting Pt: 133…136 °C
Storage Temperature: Ambient
MDL Number: MFCD01631386
CAS Number: 261762-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR