2-Chloro-p-phenylenediamine sulfate 95%

Warning

Synonyms: 2-Chloro-1,4-phenylenediammonium sulphate2-Chlorobenzene-1,4-diamine sulphate2-Chloro-1,4-phenylenediammonium sulfate2-Chlorobenzene-1,4-diamine sulfate

B24029-0B B24029-30 B24029-30 B24029-0B
AAB24029-0BEA 276.39 USD
AAB24029-0B AAB24029-30 CAAAB24029-0B CAAAB24029-30
2-Chloro-p-phenylenediamine sulfate 95%
2-Chloro-p-phenylenediamine sulphate
Formula: C₆H₉ClN₂O₄S
MW: 240.67 g/mol
Melting Pt: 251…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00013004
EINECS: 262-915-3
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR