Ethyl-3-ethoxypropionate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3-ethoxypropanoate

B24024-AU B24024-AU
AAB24024-AUEA 63.69 USD
CAAAB24024-AU AAB24024-AU
Ethyl-3-ethoxypropionate 99+%
Ethyl 3-ethoxypropionate
Formula: C₇H₁₄O₃
MW: 146.19 g/mol
Boiling Pt: 169…171 °C
Melting Pt: –100 °C
Density: 0.949
Flash Pt: 59 °C(142 °F)
Storage Temperature: Ambient
EINECS: 212-112-9
UN: 3272
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR