2'-Chloro-6'-fluoro-3'-methylacetophenone 97+%

Warning

Synonyms: 1-(2-Chloro-6-fluoro-3-methylphenyl)ethanone

B24003-06 B24003-03 B24003-03 B24003-06
AAB24003-06EA 216.27 USD
CAAAB24003-06 CAAAB24003-03 AAB24003-06 AAB24003-03
2'-Chloro-6'-fluoro-3'-methylacetophenone 97+%
2'-Chloro-6'-fluoro-3'-methylacetophenone
Formula: C₉H₈ClFO
MW: 186.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631368
CAS Number: 261762-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR