1-Naphthylacetamide 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Naphthaleneacetamide

B23986-14 B23986-22 B23986-14 B23986-36 B23986-22 B23986-36
AAB23986-14EA 32.99 USD
CAAAB23986-14 CAAAB23986-36 CAAAB23986-22 AAB23986-14 AAB23986-36 AAB23986-22
1-Naphthylacetamide 98%
1-Naphthylacetamide
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Melting Pt: 176…178 °C
Storage Temperature: Ambient
EINECS: 201-704-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR