Ethyl-3,5-bis(trifluoromethyl)benzoate 99%

Warning

B23952-06 B23952-14 B23952-03 B23952-14 B23952-06 B23952-03
AAB23952-14EA 386.28 USD
CAAAB23952-06 AAB23952-14 AAB23952-03 AAB23952-06 CAAAB23952-14 CAAAB23952-03
Ethyl-3,5-bis(trifluoromethyl)benzoate 99%
Ethyl 3,5-bis(trifluoromethyl)benzoate
Formula: C₁₁H₈F₆O₂
MW: 286.17
Storage Temperature: Ambient
MDL Number: MFCD01320684
CAS Number: 96617-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR