2-Chloro-5-(trifluoromethyl)benzyl alcohol 97+%

Warning

B23937-06 B23937-03 B23937-03 B23937-06
AAB23937-03EA 48.26 USD
AAB23937-03 AAB23937-06 CAAAB23937-06 CAAAB23937-03
2-Chloro-5-(trifluoromethyl)benzyl alcohol 97+%
2-Chloro-5-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD01631469
CAS Number: 64372-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR